2,6-Dimethoxybenzylamin
Lieferant: COMBI-BLOCKS
SS-5164-1G
COBBSS-5164-1GEA
0
EUR
COBBSS-5164-1G
2,6-Dimethoxybenzylamin
2,6-Dimethoxybenzylamin
Formel:
C₉H₁₃NO₂ Siedepunkt: 108…110 °C (1 mmHg) Schmelzpunkt: 85…86 °C |
MDL:
MFCD00052812 CAS-Nummer: 20781-22-0 |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...