3-Brom-4-fluorphenol

Lieferant: COMBI-BLOCKS

SS-5326-1G
COBBSS-5326-1GEA 21.9 EUR
COBBSS-5326-1G
3-Brom-4-fluorphenol
3-Brom-4-fluorphenol
Formel: C₆H₄BrFO
MDL: MFCD03425884
CAS-Nummer: 27407-11-0

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR