5-Amino-2-(phenoxy)benzotrifluorid

Lieferant: COMBI-BLOCKS

Synonyme: 4-Phenoxy-3-(trifluoromethyl)aniline

SS-5732-1G
COBBSS-5732-1GEA 435 EUR
COBBSS-5732-1G
5-Amino-2-(phenoxy)benzotrifluorid
5-Amino-2-(phenoxy)benzotrifluorid
Formel: C₁₃H₁₀F₃NO
Molecular Weight: 253,22 g/mol
MDL: MFCD04972663
CAS-Nummer: 267416-81-9
UN: 2811
ADR: 6.1,III

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR