2,2-Difluor-1,3-benzodioxol-4-carbonylchlorid

Lieferant: COMBI-BLOCKS

SS-5772-1G
COBBSS-5772-1GEA 137 EUR
COBBSS-5772-1G
2,2-Difluor-1,3-benzodioxol-4-carbonylchlorid
2,2-Difluor-1,3-benzodioxol-4-carbonylchlorid
Formel: C₈H₃ClF₂O₃
MDL: MFCD01631475
CAS-Nummer: 127163-51-3

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR