3-Brom-4,5-difluoranilin

Lieferant: COMBI-BLOCKS

SS-6019-1G
COBBSS-6019-1GEA 147 EUR
COBBSS-6019-1G
3-Brom-4,5-difluoranilin
3-Brom-4,5-difluoranilin
Formel: C₆H₄BrF₂N
Molecular Weight: 208,01 g/mol
MDL: MFCD08062370
CAS-Nummer: 875664-41-8
UN: 2811
ADR: 6.1,III

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR