1-Chlor-4-(4-iodphenoxy)benzol

Lieferant: COMBI-BLOCKS

Synonyme: 4-Chloro-4'-iododiphenyl ether

SS-6338-5G
COBBSS-6338-5GEA 523 EUR
COBBSS-6338-5G
1-Chlor-4-(4-iodphenoxy)benzol
1-Chloro-4-(4-iodophenoxy)benzene
Formel: C₁₂H₈ClIO
MDL: MFCD06796299
CAS-Nummer: 854257-01-5

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR