1-Fluor-4-(4-iodphenoxy)benzol

Lieferant: COMBI-BLOCKS

Synonyme: 4-Fluoro-4'-iododiphenyl ether

SS-6339-1G
COBBSS-6339-1GEA 265 EUR
COBBSS-6339-1G
1-Fluor-4-(4-iodphenoxy)benzol
1-Fluoro-4-(4-iodophenoxy)benzene
Formel: C₁₂H₈FIO
MDL: MFCD07368761
CAS-Nummer: 886762-45-4

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR