3-(Methylsulfonyl)anilin

Lieferant: COMBI-BLOCKS

SS-6364-5G
COBBSS-6364-5GEA 0 EUR
COBBSS-6364-5G
3-(Methylsulfonyl)anilin
3-(Methylsulfonyl)anilin
Formel: C₇H₉NO₂S
MDL: MFCD03764428
CAS-Nummer: 35216-39-8

Bestellen


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR