2-Brom-2'-chlor-5'-fluoracetophenon

Lieferant: COMBI-BLOCKS

Synonyme: 2-Chloro-5-fluorophenacyl bromide

SS-6378-5G
COBBSS-6378-5GEA 585 EUR
COBBSS-6378-5G
2-Brom-2'-chlor-5'-fluoracetophenon
2-Brom-2'-chlor-5'-fluoracetophenon
Formel: C₈H₅BrClFO
MDL: MFCD08059509
CAS-Nummer: 76609-34-2

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR