2-Brom-2'-fluor-4'-methoxyacetophenon

Lieferant: COMBI-BLOCKS

Synonyme: 2-Bromo-1-(2-fluoro-4-methoxyphenyl)ethanone, 2-Fluoro-4-methoxyphenacyl bromide

SS-6618-1G
COBBSS-6618-1GEA 212 EUR
COBBSS-6618-1G
2-Brom-2'-fluor-4'-methoxyacetophenon
2-Brom-2'-fluor-4'-methoxyacetophenon
Formel: C₉H₈BrFO₂
Molecular Weight: 247.0600 g/mol
MDL: MFCD03730100
CAS-Nummer: 157014-35-2

Bestellen


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR