4-(1,1,2,2-Tetrafluorethoxy)anilin

Lieferant: COMBI-BLOCKS

SS-7770-1G
COBBSS-7770-1GEA 243 EUR
COBBSS-7770-1G
4-(1,1,2,2-Tetrafluorethoxy)anilin
4-(1,1,2,2-Tetrafluorethoxy)anilin
Formel: C₈H₇F₄NO
MDL: MFCD00042377
CAS-Nummer: 713-62-2

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR