3,3-Dimethylbutyraldehyd

Lieferant: COMBI-BLOCKS

Synonyme: 3,3-Dimethylbutanal

SS-9733-5G
COBBSS-9733-5GEA 0 EUR
COBBSS-9733-5G
3,3-Dimethylbutyraldehyd
3,3-Dimethylbutyraldehyd
Formel: C₆H₁₂O
MDL: MFCD00042807
CAS-Nummer: 2987-16-8
EINECS: 221-054-3
UN: 1989
ADR: 3,III

Bestellen


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR