7-Brom-2,4-dichlorchinazolin

Lieferant: COMBI-BLOCKS

ST-3118-1G
COBBST-3118-1GEA 235 EUR
COBBST-3118-1G
7-Brom-2,4-dichlorchinazolin
7-Bromo-2,4-dichloroquinazoline
Formel: C₈H₃BrCl₂N₂
Molecular Weight: 277,93 g/mol
MDL: MFCD09954890
CAS-Nummer: 959237-68-4

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR