6-Brom-2,4-dichlorchinazolin

Lieferant: COMBI-BLOCKS

ST-4405-5G
COBBST-4405-5GEA 449 EUR
COBBST-4405-5G
6-Brom-2,4-dichlorchinazolin
6-Bromo-2,4-dichloroquinazoline
Formel: C₈H₃BrCl₂N₂
Molecular Weight: 277,93 g/mol
MDL: MFCD09744007
CAS-Nummer: 102393-82-8

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR