6-Bromisatin
Lieferant: COMBI-BLOCKS
Synonyme:
6-Bromo-1H-indole-2,3-dione, 6-Bromoindoline-2,3-dione
ST-4576-5G
COBBST-4576-5GEA
45.6
EUR
COBBST-4576-5G
6-Bromisatin
6-Bromisatin
Formel:
C₈H₄BrNO₂ Molecular Weight: 226,03 g/mol |
MDL:
MFCD01631138 CAS-Nummer: 6326-79-0 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...