1-Ethylbenzotriazol-5-carbonsäure

Lieferant: COMBI-BLOCKS

Synonyme: 1-Ethyl-1H-1,2,3-benzotriazole-5-carboxylic acid

ST-6878-1G
COBBST-6878-1GEA 413 EUR
COBBST-6878-1G
1-Ethylbenzotriazol-5-carbonsäure
1-Ethylbenzotriazole-5-carboxylic acid
Formel: C₉H₉N₃O₂
MDL: MFCD05270868
CAS-Nummer: 691363-31-2

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR