Chlor(1,5-cyclooctadien)rhodium(I) dimer

Lieferant: COMBI-BLOCKS

ST-7037-1G
COBBST-7037-1GEA 0 EUR
COBBST-7037-1G
Chlor(1,5-cyclooctadien)rhodium(I) dimer
Chlor(1,5-cyclooctadien)rhodium(I) dimer
Formel: C₁₆H₂₄Cl₂Rh₂
MDL: MFCD00012415
CAS-Nummer: 12092-47-6
EINECS: 235-157-6

Bestellen


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR