4-Amino-2-brombenzonitril
Lieferant: COMBI-BLOCKS
ST-8052-5G
COBBST-8052-5GEA
66
EUR
COBBST-8052-5G
4-Amino-2-brombenzonitril
4-Amino-2-brombenzonitril
Formel:
C₇H₅BrN₂ Molecular Weight: 197,03 g/mol |
MDL:
MFCD03695814 CAS-Nummer: 53312-82-6 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...