5-Chlor-2-methyl-7H-isoxazolo[2,3-a]pyrimidin-7-on
Lieferant: COMBI-BLOCKS
ST-8295-1G
COBBST-8295-1GEA
0
EUR
COBBST-8295-1G
5-Chlor-2-methyl-7H-isoxazolo[2,3-a]pyrimidin-7-on
5-Chlor-2-methyl-7H-isoxazolo[2,3-a]pyrimidin-7-on
Formel:
C₇H₅ClN₂O₂ |
MDL:
MFCD07366359 CAS-Nummer: 278614-92-9 |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...