3-Chlor-4-(difluormethoxy)anilin

Lieferant: COMBI-BLOCKS

ST-9283-1G
COBBST-9283-1GEA 510 EUR
COBBST-9283-1G
3-Chlor-4-(difluormethoxy)anilin
3-Chlor-4-(difluormethoxy)anilin
Formel: C₇H₆ClF₂NO
Molecular Weight: 193,58 g/mol
MDL: MFCD02724558
CAS-Nummer: 39211-55-7

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR