Kalium-1,3-dimethyluracil-5-trifluoroborat
Lieferant: COMBI-BLOCKS
TB-0008-1G
TB-0008-5G
COBBTB-0008-1GEA
420
EUR
COBBTB-0008-1G
COBBTB-0008-5G
Kalium-1,3-dimethyluracil-5-trifluoroborat
Kalium-1,3-dimethyluracil-5-trifluoroborat
Formel:
C₆H₇BF₃KN₂O₂ |
MDL:
MFCD09271782 CAS-Nummer: 1150654-77-5 |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...