Kalium-(4-trifluoroboratomethyl)thiomorpholin

Lieferant: COMBI-BLOCKS

Synonyme: Potassium trifluoro(thiomorpholinomethyl)borate

TB-0009-1G
COBBTB-0009-1GEA 276 EUR
COBBTB-0009-1G
Kalium-(4-trifluoroboratomethyl)thiomorpholin
Kalium-(4-trifluoroboratomethyl)thiomorpholin
Formel: C₅H₁₀BF₃KNS
Molecular Weight: 223,11 g/mol
MDL: MFCD10700162
CAS-Nummer: 1150654-80-0

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR