Kalium(piperidin-1-ylmethyl)trifluoroborat

Lieferant: COMBI-BLOCKS

TB-0016-1G
COBBTB-0016-1GEA 0 EUR
COBBTB-0016-1G
Kalium(piperidin-1-ylmethyl)trifluoroborat
Kalium(piperidin-1-ylmethyl)trifluoroborat
Formel: C₆H₁₂BF₃N·K
MDL: MFCD10566509
CAS-Nummer: 888711-54-4

Bestellen


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR