Kalium(pyrrolidin-1-ylmethyl)trifluoroborat

Lieferant: COMBI-BLOCKS

TB-0021-1G TB-0021-5G
COBBTB-0021-1GEA 0 EUR
COBBTB-0021-1G COBBTB-0021-5G
Kalium(pyrrolidin-1-ylmethyl)trifluoroborat
Kalium(pyrrolidin-1-ylmethyl)trifluoroborat
Formel: C₅H₁₀BF₃KN
MDL: MFCD10700161
CAS-Nummer: 888711-53-3

Bestellen


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR