Kalium(dimethylaminomethyl)trifluoroborat

Lieferant: COMBI-BLOCKS

TB-0024-1G
COBBTB-0024-1GEA 111 EUR
COBBTB-0024-1G
Kalium(dimethylaminomethyl)trifluoroborat
Kalium(dimethylaminomethyl)trifluoroborat
Formel: C₃H₈BF₃KN
MDL: MFCD10700163
CAS-Nummer: 1150655-04-1

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR