Kalium(tert-butylaminomethyl)trifluoroborat

Lieferant: COMBI-BLOCKS

TB-0040-250M
COBBTB-0040-250MEA 0 EUR
COBBTB-0040-250M
Kalium(tert-butylaminomethyl)trifluoroborat
Kalium(tert-butylaminomethyl)trifluoroborat
Formel: C₅H₁₂BF₃KN
MDL: MFCD10566510
CAS-Nummer: 1256366-05-8

Bestellen


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR