Kalium(3-(cyanmethyl)phenyl)trifluoroborat

Lieferant: COMBI-BLOCKS

TB-2445-5G TB-2445-1G
COBBTB-2445-5GEA 0 EUR
COBBTB-2445-5G COBBTB-2445-1G
Kalium(3-(cyanmethyl)phenyl)trifluoroborat
Potassium-3-(cyanomethyl)phenyltrifluoroborate
Formel: C₈H₆BF₃KN
MDL: MFCD09993212
CAS-Nummer: 1073468-32-2

Bestellen


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR