Kalium((1-phenylvinyl)trifluoroborat

Lieferant: COMBI-BLOCKS

TB-2539-1G TB-2539-25G TB-2539-5G
COBBTB-2539-1GEA 0 EUR
COBBTB-2539-1G COBBTB-2539-25G COBBTB-2539-5G
Kalium((1-phenylvinyl)trifluoroborat
Kalium((1-phenylvinyl)trifluoroborat
Formel: C₈H₇BF₃K
MDL: MFCD11052741
CAS-Nummer: 852626-70-1

Bestellen


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR