Kalium(3-methoxyphenyl)trifluoroborat

Lieferant: COMBI-BLOCKS

TB-3204-1G TB-3204-25G TB-3204-5G
COBBTB-3204-1GEA 0 EUR
COBBTB-3204-1G COBBTB-3204-25G COBBTB-3204-5G
Kalium(3-methoxyphenyl)trifluoroborat
Kalium(3-methoxyphenyl)trifluoroborat
Formel: C₇H₇BF₃KO
MDL: MFCD04039954
CAS-Nummer: 438553-44-7

Bestellen


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR