Kalium(2-(benzyloxy)phenyl)trifluoroborat

Lieferant: COMBI-BLOCKS

TB-3209-1G TB-3209-5G
COBBTB-3209-1GEA 0 EUR
COBBTB-3209-1G COBBTB-3209-5G
Kalium(2-(benzyloxy)phenyl)trifluoroborat
Kalium(2-(benzyloxy)phenyl)trifluoroborat
Formel: C₁₃H₁₁BF₃KO
MDL: MFCD04115742
CAS-Nummer: 850623-44-8

Bestellen


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR