Kalium(4-(Cbz-amino)phenyl)trifluoroborat
Lieferant: COMBI-BLOCKS
Synonyme:
Potassium 4-(benzyloxycarbonylamino)phenyltrifluoroborate
TB-3210-5G
TB-3210-1G
TB-3210-25G
COBBTB-3210-5GEA
303
EUR
COBBTB-3210-5G
COBBTB-3210-1G
COBBTB-3210-25G
Kalium(4-(Cbz-amino)phenyl)trifluoroborat
Potassium (4-(cbz-amino)phenyl)trifluoroborate
Formel:
C₁₄H₁₂BF₃KNO₂ Molecular Weight: 333.16 g/mol |
MDL:
MFCD04115743 CAS-Nummer: 850623-45-9 |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...