Kalium(4-(Cbz-amino)phenyl)trifluoroborat

Lieferant: COMBI-BLOCKS

Synonyme: Potassium 4-(benzyloxycarbonylamino)phenyltrifluoroborate

TB-3210-5G TB-3210-1G TB-3210-25G
COBBTB-3210-5GEA 303 EUR
COBBTB-3210-5G COBBTB-3210-1G COBBTB-3210-25G
Kalium(4-(Cbz-amino)phenyl)trifluoroborat
Potassium (4-(cbz-amino)phenyl)trifluoroborate
Formel: C₁₄H₁₂BF₃KNO₂
Molecular Weight: 333.16 g/mol
MDL: MFCD04115743
CAS-Nummer: 850623-45-9

Bestellen


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR