Kalium(3-cyanphenyl)trifluoroborat

Lieferant: COMBI-BLOCKS

Synonyme: (3-Cyanophenyl)trifluoroboric acid potassium salt

TB-3211-1G TB-3211-25G TB-3211-5G
COBBTB-3211-1GEA 0 EUR
COBBTB-3211-1G COBBTB-3211-25G COBBTB-3211-5G
Kalium(3-cyanphenyl)trifluoroborat
Kalium(3-cyanphenyl)trifluoroborat
Formel: C₇H₄BF₃KN
MDL: MFCD04115744
CAS-Nummer: 850623-46-0

Bestellen


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR