Kalium(2,4-dichlorphenyl)trifluoroborat

Lieferant: COMBI-BLOCKS

TB-3212-1G TB-3212-25G TB-3212-5G
COBBTB-3212-1GEA 0 EUR
COBBTB-3212-1G COBBTB-3212-25G COBBTB-3212-5G
Kalium(2,4-dichlorphenyl)trifluoroborat
Kalium(2,4-dichlorphenyl)trifluoroborat
Formel: C₆H₃BCl₂F₃K
MDL: MFCD04115745
CAS-Nummer: 192863-38-0

Bestellen


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR