Kalium(4-(benzyloxy)phenyl)trifluoroborat

Lieferant: COMBI-BLOCKS

Synonyme: Potassium (4-(benzyloxy)phenyl)trifluoroborate

TB-3213-1G TB-3213-5G
COBBTB-3213-1GEA 98.3 EUR
COBBTB-3213-1G COBBTB-3213-5G
Kalium(4-(benzyloxy)phenyl)trifluoroborat
Kalium(4-(benzyloxy)phenyl)trifluoroborat
Formel: C₁₃H₁₁BF₃KO
Molecular Weight: 290.13 g/mol
MDL: MFCD04115746
CAS-Nummer: 850623-47-1

Bestellen


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR