Kalium(4-formylphenyl)trifluoroborat
Lieferant: COMBI-BLOCKS
TB-3214-5G
TB-3214-25G
TB-3214-1G
COBBTB-3214-5GEA
0
EUR
COBBTB-3214-5G
COBBTB-3214-25G
COBBTB-3214-1G
Kalium(4-formylphenyl)trifluoroborat
Kalium(4-formylphenyl)trifluoroborat
Formel:
C₇H₅BF₃KO |
MDL:
MFCD02093980 CAS-Nummer: 374564-36-0 |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...