Kalium(4-formylphenyl)trifluoroborat

Lieferant: COMBI-BLOCKS

TB-3214-5G TB-3214-25G TB-3214-1G
COBBTB-3214-5GEA 0 EUR
COBBTB-3214-5G COBBTB-3214-25G COBBTB-3214-1G
Kalium(4-formylphenyl)trifluoroborat
Kalium(4-formylphenyl)trifluoroborat
Formel: C₇H₅BF₃KO
MDL: MFCD02093980
CAS-Nummer: 374564-36-0

Bestellen


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR