Kalium(3-chlorphenyl)trifluoroborat

Lieferant: COMBI-BLOCKS

Synonyme: (3-Chlorphenyl)trifluorborsäure Kaliumsalz

TB-3224-1G TB-3224-5G
COBBTB-3224-1GEA 0 EUR
COBBTB-3224-1G COBBTB-3224-5G
Kalium(3-chlorphenyl)trifluoroborat
Kalium(3-chlorphenyl)trifluoroborat
Formel: C₆H₄BClF₃K
MDL: MFCD04115754
CAS-Nummer: 411206-75-2

Bestellen


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR