Kaliumphenyltrifluoroborat

Lieferant: COMBI-BLOCKS

Synonyme: Phenyltrifluorborsäure Kaliumsalz

TB-3226-1G TB-3226-5G TB-3226-100G TB-3226-25G
COBBTB-3226-1GEA 26.2 EUR
COBBTB-3226-1G COBBTB-3226-5G COBBTB-3226-100G COBBTB-3226-25G
Kaliumphenyltrifluoroborat
Kaliumphenyltrifluoroborat
Formel: C₆H₅BF₃K
Molecular Weight: 184.01 g/mol
MDL: MFCD01318172
CAS-Nummer: 153766-81-5

Bestellen

TECHNISCHE DATEN

Environmentally Preferable

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR