Kalium(2-methoxyphenyl)trifluoroborat

Lieferant: COMBI-BLOCKS

Synonyme: (2-Methoxyphenyl)trifluorborsäure Kaliumsalz

TB-3227-1G TB-3227-25G TB-3227-5G
COBBTB-3227-1GEA 49.7 EUR
COBBTB-3227-1G COBBTB-3227-25G COBBTB-3227-5G
Kalium(2-methoxyphenyl)trifluoroborat
Kalium(2-methoxyphenyl)trifluoroborat
Formel: C₇H₇BF₃KO
Molecular Weight: 214.04 g/mol
MDL: MFCD04115756
CAS-Nummer: 236388-46-8

Bestellen


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR