Kalium(2-methylphenyl)trifluoroborat

Lieferant: COMBI-BLOCKS

TB-3228-1G TB-3228-25G TB-3228-5G
COBBTB-3228-1GEA 0 EUR
COBBTB-3228-1G COBBTB-3228-25G COBBTB-3228-5G
Kalium(2-methylphenyl)trifluoroborat
Kalium(2-methylphenyl)trifluoroborat
Formel: C₇H₇BF₃K
MDL: MFCD03790006
CAS-Nummer: 274257-34-0

Bestellen


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR