Kalium(4-methoxyphenyl)trifluoroborat

Lieferant: COMBI-BLOCKS

TB-3231-1G TB-3231-25G TB-3231-5G
COBBTB-3231-1GEA 0 EUR
COBBTB-3231-1G COBBTB-3231-25G COBBTB-3231-5G
Kalium(4-methoxyphenyl)trifluoroborat
Kalium(4-methoxyphenyl)trifluoroborat
Formel: C₇H₇BF₃KO
MDL: MFCD04112717
CAS-Nummer: 192863-36-8

Bestellen


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR