Kalium(4-isobutylphenyl)trifluoroborat

Lieferant: COMBI-BLOCKS

TB-3235-5G TB-3235-1G
COBBTB-3235-5GEA 0 EUR
COBBTB-3235-5G COBBTB-3235-1G
Kalium(4-isobutylphenyl)trifluoroborat
Kalium(4-isobutylphenyl)trifluoroborat
Formel: C₁₀H₁₃BF₃K
MDL: MFCD04115761
CAS-Nummer: 850623-66-4

Bestellen


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR