Kalium(3,4-dichlorphenyl)trifluoroborat

Lieferant: COMBI-BLOCKS

TB-3238-1G TB-3238-25G TB-3238-5G
COBBTB-3238-1GEA 0 EUR
COBBTB-3238-1G COBBTB-3238-25G COBBTB-3238-5G
Kalium(3,4-dichlorphenyl)trifluoroborat
Kalium(3,4-dichlorphenyl)trifluoroborat
Formel: C₆H₃BCl₂F₃K
MDL: MFCD04115763
CAS-Nummer: 850623-68-6

Bestellen


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR