Kalium(2-(methylthio)phenyl)trifluoroborat

Lieferant: COMBI-BLOCKS

TB-3250-1G TB-3250-25G TB-3250-5G
COBBTB-3250-1GEA 0 EUR
COBBTB-3250-1G COBBTB-3250-25G COBBTB-3250-5G
Kalium(2-(methylthio)phenyl)trifluoroborat
Potassium-2-(methylthio)phenyltrifluoroborate
Formel: C₇H₇BF₃KS
MDL: MFCD04115773
CAS-Nummer: 850623-77-7

Bestellen


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR