Kalium(2-fluorphenyl)trifluoroborat
Lieferant: COMBI-BLOCKS
TB-3251-5G
TB-3251-1G
TB-3251-25G
COBBTB-3251-5GEA
0
EUR
COBBTB-3251-5G
COBBTB-3251-1G
COBBTB-3251-25G
Kalium(2-fluorphenyl)trifluoroborat
Kalium(2-fluorphenyl)trifluoroborat
Formel:
C₆H₄BF₄K |
MDL:
MFCD06659937 CAS-Nummer: 166328-10-5 |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...