Kalium(3-fluorphenyl)trifluoroborat

Lieferant: COMBI-BLOCKS

TB-3252-5G TB-3252-1G TB-3252-25G
COBBTB-3252-5GEA 0 EUR
COBBTB-3252-5G COBBTB-3252-1G COBBTB-3252-25G
Kalium(3-fluorphenyl)trifluoroborat
Kalium(3-fluorphenyl)trifluoroborat
Formel: C₆H₄BF₄K
MDL: MFCD06659938
CAS-Nummer: 267006-24-6

Bestellen


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR