Kalium(4-bromphenyl)trifluoroborat

Lieferant: COMBI-BLOCKS

TB-3254-1G TB-3254-25G TB-3254-5G
COBBTB-3254-1GEA 0 EUR
COBBTB-3254-1G COBBTB-3254-25G COBBTB-3254-5G
Kalium(4-bromphenyl)trifluoroborat
Kalium(4-bromphenyl)trifluoroborat
Formel: C₆H₄BBrF₃K
MDL: MFCD02093978
CAS-Nummer: 374564-35-9

Bestellen


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR