Kalium(2-naphthyl)trifluoroborat

Lieferant: COMBI-BLOCKS

TB-3256-1G TB-3256-25G TB-3256-5G
COBBTB-3256-1GEA 0 EUR
COBBTB-3256-1G COBBTB-3256-25G COBBTB-3256-5G
Kalium(2-naphthyl)trifluoroborat
Kalium(2-naphthyl)trifluoroborat
Formel: C₁₀H₇BF₃K
MDL: MFCD06659941
CAS-Nummer: 668984-08-5

Bestellen


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR