3'-Fluor-4'-hydroxybiphenyl-4-carbonsäure

Lieferant: COMBI-BLOCKS

YA-1074-5G
COBBYA-1074-5GEA 2040 EUR
COBBYA-1074-5G
3'-Fluor-4'-hydroxybiphenyl-4-carbonsäure
3'-Fluor-4'-hydroxybiphenyl-4-carbonsäure
Formel: C₁₃H₉FO₃
MDL: MFCD00994624
CAS-Nummer: 106291-26-3

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR